Alfa Aesar™ N-(Hydroxymethyl)acetamide, 98+%

Manufacturer: Alfa Aesar™ 3167014

Quantity 25g

 View more versions of this product

Catalog No. AA3167014

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 625-51-4
    Formula Weight 89.1
    MDL Number MFCD00014417
    Chemical Name or Material N-(Hydroxymethyl)acetamide

    Specifications

    Formula Weight 89.1
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00014417
    CAS 625-51-4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H332-H319
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    Melting Point 55° to 56°C
    Quantity 25g
    Physical Form Crystalline
    Chemical Name or Material N-(Hydroxymethyl)acetamide
    Assay ≥98%

    GHS H Statement
    H311-H332-H319
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit