Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Hydroxymethyl)benzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6282-02-6
97.5
100.0
98%
C8H9NO2
C6H5CONHCH2OH
MFCD00014415
n-hydroxymethyl benzamide,benzamide, n-hydroxymethyl,n-methylolbenzamide,acmc-20akah,pubchem12540,enamine_001590,n-hydroxymetyl-benzamide,benzamide,n-hydroxymethyl,bis 5-nitropyridin-4-yl sulfane
N-(Hydroxymethyl)benzamide, 98%
6282-02-6
97.5
100.0
98%
C8H9NO2
C6H5CONHCH2OH
MFCD00014415
n-hydroxymethyl benzamide,benzamide, n-hydroxymethyl,n-methylolbenzamide,acmc-20akah,pubchem12540,enamine_001590,n-hydroxymetyl-benzamide,benzamide,n-hydroxymethyl,bis 5-nitropyridin-4-yl sulfane
N-(Hydroxymethyl)benzamide, 98%
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Quantity 25g
InChI Key UOUBPDZUBVJZOQ-UHFFFAOYSA-N
Molecular Weight (g/mol) 151.17
PubChem CID 80493
Formula Weight 151.16
Physical Form Crystalline Powder
Percent Purity ≥97.5%

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.