Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Hydroxymethyl)phthalimide 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
118-29-6
C9H7NO3
177.16
MFCD00005899
MNSGOOCAMMSKGI-UHFFFAOYSA-N
n-hydroxymethyl phthalimide,n-hydroxymethylphthalimide,2-hydroxymethyl isoindoline-1,3-dione,phthalimidomethanol,phthalimidomethyl alcohol,n-methylolphthalimide,oxymethyl phthalimide,1h-isoindole-1,3 2h-dione, 2-hydroxymethyl,hydroxymethylphthalimide,methanol, phthalimido
8354
CHEBI:38816
2-(hydroxymethyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CO
118-29-6
C9H7NO3
177.16
MFCD00005899
MNSGOOCAMMSKGI-UHFFFAOYSA-N
n-hydroxymethyl phthalimide,n-hydroxymethylphthalimide,2-hydroxymethyl isoindoline-1,3-dione,phthalimidomethanol,phthalimidomethyl alcohol,n-methylolphthalimide,oxymethyl phthalimide,1h-isoindole-1,3 2h-dione, 2-hydroxymethyl,hydroxymethylphthalimide,methanol, phthalimido
8354
CHEBI:38816
2-(hydroxymethyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CO
Sulfated Ash 0.1% max.
Quantity 25g
Melting Point 140°C to 145°C
Infrared Spectrum Authentic
Packaging Glass bottle
Loss on Drying 2% max.
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 21, 475
Fieser 01,484; 08,253
Formula Weight 177.16
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material N-(Hydroxymethyl)phthalimide, 97%
Show More Show Less

EINECSNumber : 204-241-4

RTECSNumber : TI5270000

TSCA : TSCA