Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxyurethane, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
589-41-3
C3H7NO3
105.093
VGEWEGHHYWGXGG-UHFFFAOYSA-N
n-hydroxyurethane,ethyl hydroxycarbamate,n-hydroxyurethan,hydroxyurethane,hydroxycarbamic acid ethyl ester,n-carbethoxyhydroxylamine,n-hydroxy ethyl carbamate,hydroxyurethan,carbamic acid, hydroxy-, ethyl ester,ethyl-n-hydroxycarbamate
11510
ethyl N-hydroxycarbamate
CCOC(=O)NO
589-41-3
C3H7NO3
105.093
VGEWEGHHYWGXGG-UHFFFAOYSA-N
n-hydroxyurethane,ethyl hydroxycarbamate,n-hydroxyurethan,hydroxyurethane,hydroxycarbamic acid ethyl ester,n-carbethoxyhydroxylamine,n-hydroxy ethyl carbamate,hydroxyurethan,carbamic acid, hydroxy-, ethyl ester,ethyl-n-hydroxycarbamate
11510
ethyl N-hydroxycarbamate
CCOC(=O)NO
Quantity 25g
Elemental Analysis 30.86 to 37.72% C, 12.00 to 14.66% N
Color Brown to Yellow
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula HONHCO2C2H5
Fieser 17,150; 01,1228; 03,308; 06,622
Formula Weight 105.09
Physical Form Liquid
Chemical Name or Material N-Hydroxyurethane
Show More Show Less