Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Iodosaccharin 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
86340-94-5
C7H4INO3S
309.077
MFCD09037939
XQKQROJYWLWDMP-UHFFFAOYSA-N
n-iodosaccharin,unii-11wjr74obi,11wjr74obi,1,2-benzisothiazol-3 2h-one, 2-iodo-, 1,1-dioxide,2-iodo-1,2-benzisothiazol-3 2h-one 1,1-dioxide,n-iodosaccharin mi,2-iodo-1??,2-benzothiazole-1,1,3-trione,n-yodosaccharin,2-iodobenzo d isothiazol-3 2h-one 1,1-dioxide
8063362
2-iodo-1,1-dioxo-1,2-benzothiazol-3-one
C1=CC=C2C(=C1)C(=O)N(S2(=O)=O)I
86340-94-5
C7H4INO3S
309.077
MFCD09037939
XQKQROJYWLWDMP-UHFFFAOYSA-N
n-iodosaccharin,unii-11wjr74obi,11wjr74obi,1,2-benzisothiazol-3 2h-one, 2-iodo-, 1,1-dioxide,2-iodo-1,2-benzisothiazol-3 2h-one 1,1-dioxide,n-iodosaccharin mi,2-iodo-1??,2-benzothiazole-1,1,3-trione,n-yodosaccharin,2-iodobenzo d isothiazol-3 2h-one 1,1-dioxide
8063362
2-iodo-1,1-dioxo-1,2-benzothiazol-3-one
C1=CC=C2C(=C1)C(=O)N(S2(=O)=O)I
C-X bond formation reagent
Packaging Glass bottle
Color Beige
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Merck Index 15, 5086
Formula Weight 309.08
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material N-Iodosaccharin
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.