Alfa Aesar™ N-Isobutyryl-2'-deoxyguanosine, 97%

Manufacturer: Alfa Aesar™ H52730MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52730MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142554-22-1
    Molecular Formula C14H19N5O5
    Formula Weight 337.34
    MDL Number MFCD02682949
    Chemical Name or Material N-Isobutyryl-2'-deoxyguanosine

    Specifications

    Formula Weight 337.34
    Molecular Formula C14H19N5O5
    MDL Number MFCD02682949
    CAS 142554-22-1
    Quantity 250mg
    Chemical Name or Material N-Isobutyryl-2'-deoxyguanosine
    Assay 97%

    Description

    Applications
    N-Isobutyryl-2 ft.-deoxyguanosine, is used as pharmaceutical intermediate.

    Solubility
    It is water soluble.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit