Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropyl-1-piperazineacetamide, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
39890-42-1
C9H19N3O
185.271
MFCD00005969
FFOHTSTWXWJGQR-UHFFFAOYSA-N
n-isopropyl-1-piperazineacetamide,n-isopropyl-2-piperazin-1-yl acetamide,n-isopropyl-2-piperazin-1-ylacetamide,2-piperazin-1-yl-n-propan-2-yl acetamide,1-piperazineacetamide, n-1-methylethyl,n-methylethyl-2-piperazinylacetamide,n-isopropyl-2-piperazinoacetamide,2-piperazinyl-n-isopropylacetamide,n-isopropyl-1-piperazine acetamide,piperazinoacetic acid isopropylamide
100826
2-piperazin-1-yl-N-propan-2-ylacetamide
CC(C)NC(=O)CN1CCNCC1
39890-42-1
C9H19N3O
185.271
MFCD00005969
FFOHTSTWXWJGQR-UHFFFAOYSA-N
n-isopropyl-1-piperazineacetamide,n-isopropyl-2-piperazin-1-yl acetamide,n-isopropyl-2-piperazin-1-ylacetamide,2-piperazin-1-yl-n-propan-2-yl acetamide,1-piperazineacetamide, n-1-methylethyl,n-methylethyl-2-piperazinylacetamide,n-isopropyl-2-piperazinoacetamide,2-piperazinyl-n-isopropylacetamide,n-isopropyl-1-piperazine acetamide,piperazinoacetic acid isopropylamide
100826
2-piperazin-1-yl-N-propan-2-ylacetamide
CC(C)NC(=O)CN1CCNCC1
Infrared Spectrum Authentic
Melting Point 82°C to 88°C
Color Yellow
Packaging Glass bottle
Quantity 25g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 185.27
Physical Form Crystals and Chunks
Percent Purity ≥94%
Chemical Name or Material N-Isopropyl-1-piperazineacetamide, 95%
Show More Show Less

EINECSNumber : 254-676-9