Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Isopropyl-2-chloroacetamide, 97%

Click to view available options
Quantity:
1g
5g
2895-21-8
C5H10ClNO
135.591
MFCD00068034
GYPNJSBBOATUPK-UHFFFAOYSA-N
2-chloro-n-isopropylacetamide,n-isopropyl-2-chloroacetamide,2-chloro-n-1-methylethyl acetamide,n-isopropyl chloroacetamide,2-chloro-n-propan-2-yl acetamide,acetamide, 2-chloro-n-1-methylethyl,n-chloroacetyl isopropylamine,2-chloro-n-methylethyl acetamide,acmc-1cpp2,2-chloro-n-isopropyl acetamide
76171
2-chloro-N-propan-2-ylacetamide
CC(C)NC(=O)CCl
2895-21-8
C5H10ClNO
135.591
MFCD00068034
GYPNJSBBOATUPK-UHFFFAOYSA-N
2-chloro-n-isopropylacetamide,n-isopropyl-2-chloroacetamide,2-chloro-n-1-methylethyl acetamide,n-isopropyl chloroacetamide,2-chloro-n-propan-2-yl acetamide,acetamide, 2-chloro-n-1-methylethyl,n-chloroacetyl isopropylamine,2-chloro-n-methylethyl acetamide,acmc-1cpp2,2-chloro-n-isopropyl acetamide
76171
2-chloro-N-propan-2-ylacetamide
CC(C)NC(=O)CCl
Quantity 5g
Melting Point 58°C to 60°C
Beilstein 1745034
Formula Weight 135.59
Percent Purity 97%
Chemical Name or Material N-Isopropyl-2-chloroacetamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning