Alfa Aesar™ N-Isopropyl-N'-phenyl-p-phenylenediamine, 95%

Manufacturer: Alfa Aesar™ B2537830

Quantity 250g

 View more versions of this product

Catalog No. AAB2537830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 101-72-4
    Molecular Formula C15H18N2
    Formula Weight 226.32
    MDL Number MFCD00026348
    UN Number UN3077
    Synonym 4-(Isopropylamino)diphenylamine
    Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine

    Specifications

    Formula Weight 226.32
    Health Hazard 3 GHS P Statement
    P280g-P262-P273-P501a
    Wear protective gloves.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C15H18N2
    MDL Number MFCD00026348
    Flash Point 172°C (342°F)
    Quantity 250g
    CAS 101-72-4
    Synonym 4-(Isopropylamino)diphenylamine
    Health Hazard 1 Warning
    RTECS Number ST2650000
    TSCA TSCA
    UN Number UN3077
    Health Hazard 2 GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Boiling Point 161°C (1mmHg)
    Melting Point 74° to 80°C
    Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine
    Assay 95%

    GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P280g-P262-P273-P501a
    Wear protective gloves.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit