Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Isopropyl-N'-phenyl-p-phenylenediamine, 95%

Click to view available options
Quantity:
250g
1kg
101-72-4
C15H18N2
226.323
MFCD00026348
OUBMGJOQLXMSNT-UHFFFAOYSA-N
ippd,n-isopropyl-n'-phenyl-p-phenylenediamine,n-isopropyl-n'-phenyl-1,4-phenylenediamine,cyzone,4-anilino-n-isopropylaniline,antigen 3c,antioxidant ip,santoflex ip,diaphen fp,cyzone ip
7573
CHEBI:63569
1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2
101-72-4
C15H18N2
226.323
MFCD00026348
OUBMGJOQLXMSNT-UHFFFAOYSA-N
ippd,n-isopropyl-n'-phenyl-p-phenylenediamine,n-isopropyl-n'-phenyl-1,4-phenylenediamine,cyzone,4-anilino-n-isopropylaniline,antigen 3c,antioxidant ip,santoflex ip,diaphen fp,cyzone ip
7573
CHEBI:63569
1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2
Boiling Point 161°C (1mmHg)
Melting Point 74°C to 80°C
Quantity 1kg
Flash Point 172°C (342°F)
UN Number UN3077
Formula Weight 226.32
Percent Purity 95%
Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P280g-P262-P273-P501a
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ST2650000

TSCA : TSCA