Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropylcyclohexylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1195-42-2
C9H19N
141.26
MFCD00003831
UYYCVBASZNFFRX-UHFFFAOYSA-N
n-isopropylcyclohexylamine,cyclohexylisopropylamine,n-isopropylcyclohexanamine,cyclohexylamine, n-isopropyl,isopropylcyclohexylamine,cyclohexanamine, n-1-methylethyl,n-cyclohexylisopropylamine,n-cyclohexyl-n-isopropylamine,n-propan-2-yl cyclohexanamine,n-isopropyl cyclohexylamine
62386
N-propan-2-ylcyclohexanamine
CC(C)NC1CCCCC1
1195-42-2
C9H19N
141.26
MFCD00003831
UYYCVBASZNFFRX-UHFFFAOYSA-N
n-isopropylcyclohexylamine,cyclohexylisopropylamine,n-isopropylcyclohexanamine,cyclohexylamine, n-isopropyl,isopropylcyclohexylamine,cyclohexanamine, n-1-methylethyl,n-cyclohexylisopropylamine,n-cyclohexyl-n-isopropylamine,n-propan-2-yl cyclohexanamine,n-isopropyl cyclohexylamine
62386
N-propan-2-ylcyclohexanamine
CC(C)NC1CCCCC1
Boiling Point 175°C
Flash Point 33°C
Specific Gravity 0.85
Refractive Index 1.4470 to 1.449
Density 0.8500g/mL
Quantity 100g
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H11NHCH(CH3)2
Formula Weight 141.26
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Isopropylcyclohexylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-798-5