Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropylethylenediamine 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
19522-67-9
C5H14N2
102.181
MFCD00008164
KDRUIMNNZBMLJR-UHFFFAOYSA-N
n-isopropylethylenediamine,2-isopropylaminoethylamine,isopropylaminoethylamine,1,2-ethanediamine, n-1-methylethyl,ethylenediamine, n-isopropyl,n-isopropylethane-1,2-diamine,2-aminoethyl propan-2-yl amine,1,2-ethanediamine, n1-1-methylethyl,2-aminoethyl isopropyl amine,n1-isopropylethane-1,2-diamine
88098
CHEBI:84288
N'-propan-2-ylethane-1,2-diamine
CC(C)NCCN
19522-67-9
C5H14N2
102.181
MFCD00008164
KDRUIMNNZBMLJR-UHFFFAOYSA-N
n-isopropylethylenediamine,2-isopropylaminoethylamine,isopropylaminoethylamine,1,2-ethanediamine, n-1-methylethyl,ethylenediamine, n-isopropyl,n-isopropylethane-1,2-diamine,2-aminoethyl propan-2-yl amine,1,2-ethanediamine, n1-1-methylethyl,2-aminoethyl isopropyl amine,n1-isopropylethane-1,2-diamine
88098
CHEBI:84288
N'-propan-2-ylethane-1,2-diamine
CC(C)NCCN
Quantity 10g
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4359 to 1.4379 (20°C, 589nm)
Density 0.81g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2CHNHCH2CH2NH2
Formula Weight 102.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material N-Isopropylethylenediamine, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger