Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-4323707
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
4747-21-1
C4H11N
73.14
XHFGWHUWQXTGAT-UHFFFAOYSA-N
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
78485
N-methylpropan-2-amine
CC(C)NC
4747-21-1
C4H11N
73.14
XHFGWHUWQXTGAT-UHFFFAOYSA-N
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
78485
N-methylpropan-2-amine
CC(C)NC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7020g/mL
Boiling Point 50°C to 53°C
Flash Point -32°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Linear Formula CH3NHCH(CH3)2
Quantity 10 mL
Specific Gravity 0.702
Formula Weight 73.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Isopropylmethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
EINECSNumber 225-266-7

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit