Alfa Aesar™ N-Isopropylpropylamine, 96%

Manufacturer: Alfa Aesar™ H50256MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50256MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21968-17-2
    Molecular Formula C6H15N
    Linear Formula CH3CH2CH2NHCH(CH3)2
    Formula Weight 101.19
    MDL Number MFCD00027095
    UN Number UN2733
    Chemical Name or Material N-Isopropylpropylamine

    Specifications

    Formula Weight 101.19
    Molecular Formula C6H15N
    Linear Formula CH3CH2CH2NHCH(CH3)2
    CAS 21968-17-2
    UN Number UN2733
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H224-H314-H318
    Extremely flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 250mg
    MDL Number MFCD00027095
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Chemical Name or Material N-Isopropylpropylamine
    Assay 96%

    Description

    Applications
    N-Isopropylpropylamine is used as an active pharmaceutical ingredients.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H224-H314-H318
    Extremely flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit