Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroylsarcosine sodium salt, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
137-16-6
C15H28NO3·Na
293.39
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sarkosyl nl,sodium lauroyl sarcosinate,n-lauroylsarcosine sodium salt,sodium n-lauroylsarcosinate,sodium lauroylsarcosinate,sarcosyl nl,maprosyl 30,compound 105,gardol,hamposyl l-30
23668817
sodium;2-[dodecanoyl(methyl)amino]acetate
CCCCCCCCCCCC(=O)N(C)CC(=O)[O-].[Na+]
137-16-6
C15H28NO3·Na
293.39
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sarkosyl nl,sodium lauroyl sarcosinate,n-lauroylsarcosine sodium salt,sodium n-lauroylsarcosinate,sodium lauroylsarcosinate,sarcosyl nl,maprosyl 30,compound 105,gardol,hamposyl l-30
23668817
sodium;2-[dodecanoyl(methyl)amino]acetate
CCCCCCCCCCCC(=O)N(C)CC(=O)[O-].[Na+]
pH 7.0 to 9.0
Loss on Drying 7% max. ()
Quantity 1kg
Water 7.9% max.
Infrared Spectrum Authentic
Color White
Appearance (Almost clear in water)
Packaging Glass bottle
Melting Point 45.0°C to 47.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula NaCO2CH2N(CH3)CO(CH2)10CH3
Merck Index 15,4401
Solubility Information (10% in water) Almost clear colorless to light yellow
Formula Weight 293.39
Physical Form Powder
Percent Purity 95%
Chemical Name or Material N-Lauroylsarcosine sodium salt
Show More Show Less

GHS H Statement:

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 205-281-5