Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Linoleoylglycine, 98%

Click to view available options
Quantity:
5mg
10mg
2768-02-7
C5H12O3Si
148.23
MFCD11977729
NKSJNEHGWDZZQF-UHFFFAOYSA-N
vinyltrimethoxysilane,trimethoxyvinylsilane,silane, ethenyltrimethoxy,trimethoxy vinyl silane,trimethoxysilyl ethene,vinyl trimethoxy silane,silane, trimethoxyvinyl,trimethoxysilyl ethylene,vtmo,vts-m
76004
ethenyl(trimethoxy)silane
CO[Si](C=C)(OC)OC
2768-02-7
C5H12O3Si
148.23
MFCD11977729
NKSJNEHGWDZZQF-UHFFFAOYSA-N
vinyltrimethoxysilane,trimethoxyvinylsilane,silane, ethenyltrimethoxy,trimethoxy vinyl silane,trimethoxysilyl ethene,vinyl trimethoxy silane,silane, trimethoxyvinyl,trimethoxysilyl ethylene,vtmo,vts-m
76004
ethenyl(trimethoxy)silane
CO[Si](C=C)(OC)OC
Formula Weight 337.5
Physical Form Solid
Percent Purity 98%
Quantity 5mg
Chemical Name or Material N-Linoleoylglycine

RTECSNumber : MC0596320

storageNote1 : -30° to -10°C