Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methoxy-N-methylacetamide, 98%

Click to view available options
Quantity:
5g
25g
100g
78191-00-1
C4H9NO2
103.121
MFCD00060098
OYVXVLSZQHSNDK-UHFFFAOYSA-N
n-methyl-n-methoxyacetamide,acetamide, n-methoxy-n-methyl,n-methoxyl-n-methylacetamide,n-methoxy-n-methyl-acetamide,n-methoxy-n-methylacetamid,pubchem12775,acmc-209pdo,n-methoxy-n-methylacetamde,n-methoxy n-methylacetamide,n-methoxy-n-methyl acetamide
537505
N-methoxy-N-methylacetamide
CC(=O)N(C)OC
78191-00-1
C4H9NO2
103.121
MFCD00060098
OYVXVLSZQHSNDK-UHFFFAOYSA-N
n-methyl-n-methoxyacetamide,acetamide, n-methoxy-n-methyl,n-methoxyl-n-methylacetamide,n-methoxy-n-methyl-acetamide,n-methoxy-n-methylacetamid,pubchem12775,acmc-209pdo,n-methoxy-n-methylacetamde,n-methoxy n-methylacetamide,n-methoxy-n-methyl acetamide
537505
N-methoxy-N-methylacetamide
CC(=O)N(C)OC
Color Colorless
Boiling Point 152°C
Density 0.970g/mL
Quantity 100g
Refractive Index 1.4260
UN Number UN1993
Beilstein 3600878
Formula Weight 103.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material N-Methoxy-N-methylacetamide
Show More Show Less

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.