Alfa Aesar™ N-Methoxy-N-methylbenzamide, 98%

Manufacturer: Alfa Aesar™ H5425314

Quantity 25g

 View more versions of this product

Catalog No. AAH5425314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6919-61-5
    Molecular Formula C9H11NO2
    Formula Weight 165.19
    MDL Number MFCD00075320
    Chemical Name or Material N-Methoxy-N-methylbenzamide

    Specifications

    Formula Weight 165.19
    Quantity 25g
    Boiling Point 70°C
    Flash Point 113°C (235°F)
    Molecular Formula C9H11NO2
    MDL Number MFCD00075320
    CAS 6919-61-5
    Chemical Name or Material N-Methoxy-N-methylbenzamide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit