Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methoxy-N-methylcarbamoyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-3213002
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
30289-28-2
C3H6ClNO2
123.54
MFCD06658962
SHQJHXLWZDMQEJ-UHFFFAOYSA-N
acmc-209he9, methylmethoxycarbamoyl chloride, methoxy methyl carbamic chloride, n-methoxy-n-methylcarbamic chloride, n-methoxy-n-methylcarbamyl chloride, n-methyl-n-methoxycarbamyl chloride, carbamic chloride,n-methoxy-n-methyl, n-methoxy-n-methylcarbamic acid chloride
2734926
N-methoxy-N-methylcarbamoyl chloride
CON(C)C(Cl)=O
30289-28-2
C3H6ClNO2
123.54
MFCD06658962
SHQJHXLWZDMQEJ-UHFFFAOYSA-N
acmc-209he9, methylmethoxycarbamoyl chloride, methoxy methyl carbamic chloride, n-methoxy-n-methylcarbamic chloride, n-methoxy-n-methylcarbamyl chloride, n-methyl-n-methoxycarbamyl chloride, carbamic chloride,n-methoxy-n-methyl, n-methoxy-n-methylcarbamic acid chloride
2734926
N-methoxy-N-methylcarbamoyl chloride
CON(C)C(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4420 to 1.444
Linear Formula CH3ON(CH3)COCl
Quantity 5 g
Formula Weight 123.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylcarbamoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit