Alfa Aesar™ N-Methoxycarbonyl-L-tert-leucine, 98%

Manufacturer: Alfa Aesar™ H3514806

Quantity 5g

 View more versions of this product

Catalog No. AAH3514806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 162537-11-3
    Molecular Formula C8H15NO4
    Formula Weight 189.21
    MDL Number MFCD07778455
    Synonym N-Moc-L-tert-leucine
    Chemical Name or Material N-Methoxycarbonyl-L-tert-leucine

    Specifications

    Formula Weight 189.21
    Quantity 5g
    MDL Number MFCD07778455
    Synonym N-Moc-L-tert-leucine
    Recommended Storage 2° to 8°C
    Melting Point 107° to 110°C
    Molecular Formula C8H15NO4
    CAS 162537-11-3
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material N-Methoxycarbonyl-L-tert-leucine
    Assay 98%

    Description

    Applications
    The coupling of the two key intermediates, N-(methoxycarbonyl)-l-tert-leucine acylated benzyl hydrazine and chloromethyl ketone, via an S N 2 reaction furnished the amino ketone in high yield under our optimized conditions in practical synthesis of the HIV Protease Inhibitor Atazanavir via a Highly diastereoselective Reduction Approach.

    Solubility
    Soluble in ethyl acetate and methanol.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit