Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
93102-05-7
C13H23NOSi
237.42
MFCD00674005
RPZAAFUKDPKTKP-UHFFFAOYSA-N
n-methoxymethyl-n-trimethylsilylmethyl benzylamine,n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine,benzyl methoxymethyl trimethylsilyl methyl amine,benzyl-methoxymethyl-trimethylsilanylmethyl-amine,n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine,n-benzyl-n-methoxymethyl trimethylsilylmethylamine,n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine,n-methoxymethyl-n-trimethylsilyl methyl phenyl,n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
353442
N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)methanamine
COCN(CC1=CC=CC=C1)C[Si](C)(C)C
93102-05-7
C13H23NOSi
237.42
MFCD00674005
RPZAAFUKDPKTKP-UHFFFAOYSA-N
n-methoxymethyl-n-trimethylsilylmethyl benzylamine,n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine,benzyl methoxymethyl trimethylsilyl methyl amine,benzyl-methoxymethyl-trimethylsilanylmethyl-amine,n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine,n-benzyl-n-methoxymethyl trimethylsilylmethylamine,n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine,n-methoxymethyl-n-trimethylsilyl methyl phenyl,n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
353442
N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)methanamine
COCN(CC1=CC=CC=C1)C[Si](C)(C)C
Boiling Point 76°C (0.3mmHg)
Flash Point 66°C
Specific Gravity 0.928
Refractive Index 1.4915 to 1.4935
Color Colorless to Yellow
Infrared Spectrum Authentic
Density 0.9280g/mL
Elemental Analysis 95% min.
Packaging Glass bottle
Quantity 5g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula C6H5CH2N(CH2OCH3)CH2Si(CH3)3
Formula Weight 237.42
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning