Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-1,3-propanediamine, 99%

Click to view available options
Quantity:
100g
500g
6291-84-5
C4H12N2
88.154
MFCD00008209
QHJABUZHRJTCAR-UHFFFAOYSA-N
n-methyl-1,3-propanediamine,n-methyl-1,3-diaminopropane,3-methylamino propylamine,1,3-propanediamine, n-methyl,n-methylpropane-1,3-diamine,3-aminopropyl methylamine,n-methyltrimethylenediamine,3-aminopropylmethylamine,n-methyl-1,3-propylenediamine,1-amino-3-methylamino propane
80511
N'-methylpropane-1,3-diamine
CNCCCN
6291-84-5
C4H12N2
88.154
MFCD00008209
QHJABUZHRJTCAR-UHFFFAOYSA-N
n-methyl-1,3-propanediamine,n-methyl-1,3-diaminopropane,3-methylamino propylamine,1,3-propanediamine, n-methyl,n-methylpropane-1,3-diamine,3-aminopropyl methylamine,n-methyltrimethylenediamine,3-aminopropylmethylamine,n-methyl-1,3-propylenediamine,1-amino-3-methylamino propane
80511
N'-methylpropane-1,3-diamine
CNCCCN
Quantity 500g
Melting Point -72°C
Flash Point 35°C (96°F)
Boiling Point 139°C to 141°C
UN Number UN2734
Beilstein 878143
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material N-Methyl-1,3-propanediamine

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

RTECSNumber : TX8242500

TSCA : TSCA