Alfa Aesar™ N-Methyl-1,3-propanediamine, 99%

Manufacturer: Alfa Aesar™ B2331922

Quantity 100g

 View more versions of this product

Catalog No. AAB2331922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6291-84-5
    Molecular Formula CH3NH(CH2)3NH2
    Formula Weight 88.15
    MDL Number MFCD00008209
    UN Number UN2734
    Beilstein 878143
    Synonym 3-Methylamino-1-propylamine; N-Methyl-1,3-diaminopropane
    Chemical Name or Material N-Methyl-1,3-propanediamine

    Specifications

    Formula Weight 88.15
    TSCA TSCA
    Melting Point −72°C
    Boiling Point 139° to 141°C
    Quantity 100g
    Flash Point 35°C (96°F)
    Molecular Formula CH3NH(CH2)3NH2
    CAS 6291-84-5
    UN Number UN2734
    Beilstein 878143
    Synonym 3-Methylamino-1-propylamine; N-Methyl-1,3-diaminopropane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H226
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    RTECS Number TX8242500
    MDL Number MFCD00008209
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
    Chemical Name or Material N-Methyl-1,3-propanediamine
    Assay 99%

    GHS H Statement
    H301-H314-H318-H226
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit