Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-D-alanine, 98%

Click to view available options
Quantity:
5g
25g
29475-64-7
C4H9NO2
103.121
MFCD00063135
GDFAOVXKHJXLEI-GSVOUGTGSA-N
n-methyl-d-alanine,r-2-methylamino propanoic acid,h-d-meala-oh,d-alanine, n-methyl,2r-2-methylamino propanoic acid,h-n-me-d-ala-oh,2r-2-methylamino propanoic acid hydrochloride,n-methyl-d-ala,ambotzhaa1193,ksc201s8d
92973
(2R)-2-(methylamino)propanoic acid
CC(C(=O)O)NC
29475-64-7
C4H9NO2
103.121
MFCD00063135
GDFAOVXKHJXLEI-GSVOUGTGSA-N
n-methyl-d-alanine,r-2-methylamino propanoic acid,h-d-meala-oh,d-alanine, n-methyl,2r-2-methylamino propanoic acid,h-n-me-d-ala-oh,2r-2-methylamino propanoic acid hydrochloride,n-methyl-d-ala,ambotzhaa1193,ksc201s8d
92973
(2R)-2-(methylamino)propanoic acid
CC(C(=O)O)NC
Beilstein 1720923
Formula Weight 103.12
Physical Form Powder
Percent Purity 98%
Color White
Quantity 25g
Chemical Name or Material N-Methyl-D-alanine

storageNote1 : -30° to -10°C