Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-D-phenylalanine, 97%

Click to view available options
Quantity:
250mg
1g
5g
56564-52-4
C10H13NO2
179.219
MFCD00069560
SCIFESDRCALIIM-SECBINFHSA-N
r-2-methylamino-3-phenylpropanoic acid,n-methyl-d-phenylalanine,d-phenylalanine, n-methyl,2r-2-methylamino-3-phenylpropanoic acid,h-d-mephe-oh hcl,ambotzhaa1199,l-phenylalanine, n-methyl
6427037
(2R)-2-(methylamino)-3-phenylpropanoic acid
CNC(CC1=CC=CC=C1)C(=O)O
56564-52-4
C10H13NO2
179.219
MFCD00069560
SCIFESDRCALIIM-SECBINFHSA-N
r-2-methylamino-3-phenylpropanoic acid,n-methyl-d-phenylalanine,d-phenylalanine, n-methyl,2r-2-methylamino-3-phenylpropanoic acid,h-d-mephe-oh hcl,ambotzhaa1199,l-phenylalanine, n-methyl
6427037
(2R)-2-(methylamino)-3-phenylpropanoic acid
CNC(CC1=CC=CC=C1)C(=O)O
Beilstein 3199933
Formula Weight 179.22
Physical Form Solid
Percent Purity 97%
Quantity 1g
Chemical Name or Material N-Methyl-D-phenylalanine