Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-DL-alanine, 98%

Click to view available options
Quantity:
25g
100g
500g
600-21-5
C4H9NO2
103.121
MFCD00063136
GDFAOVXKHJXLEI-UHFFFAOYSA-N
n-methyl-dl-alanine,2-methylamino propanoic acid,h-n-me-dl-ala-oh,n-alpha-methyl-dl-alanine,n-me-dl-ala-oh,alanine, n-methyl-, l,dl-n-methylalanine,me-dl-ala-oh,n-methyl cdl-alanine,n-,a-methyl-dl-alanine
4377
2-(methylamino)propanoic acid
CC(C(=O)O)NC
600-21-5
C4H9NO2
103.121
MFCD00063136
GDFAOVXKHJXLEI-UHFFFAOYSA-N
n-methyl-dl-alanine,2-methylamino propanoic acid,h-n-me-dl-ala-oh,n-alpha-methyl-dl-alanine,n-me-dl-ala-oh,alanine, n-methyl-, l,dl-n-methylalanine,me-dl-ala-oh,n-methyl cdl-alanine,n-,a-methyl-dl-alanine
4377
2-(methylamino)propanoic acid
CC(C(=O)O)NC
Formula Weight 103.12
Physical Form Crystalline Powder
Percent Purity 98%
Color White
Quantity 25g
Chemical Name or Material N-Methyl-DL-alanine