Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-L-phenylalanine benzyl ester p-toluenesulfonate, 95%

Click to view available options
Quantity:
250mg
1g
40298-25-7
C24H27NO5S
441.542
MFCD00238321
TUDJZSODAYRPLC-NTISSMGPSA-N
n-me-phe-obzl p-tosylate,n-me-phe-obzl tosoh,4-methylbenzene-1-sulfonic acid-benzyl n-methyl-l-phenylalaninate 1/1,benzyl 2s-2-methylamino-3-phenylpropanoate; para-toluene sulfonate
56777189
benzyl (2S)-2-(methylamino)-3-phenylpropanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.CNC(CC1=CC=CC=C1)C(=O)OCC2=CC=CC=C2
40298-25-7
C24H27NO5S
441.542
MFCD00238321
TUDJZSODAYRPLC-NTISSMGPSA-N
n-me-phe-obzl p-tosylate,n-me-phe-obzl tosoh,4-methylbenzene-1-sulfonic acid-benzyl n-methyl-l-phenylalaninate 1/1,benzyl 2s-2-methylamino-3-phenylpropanoate; para-toluene sulfonate
56777189
benzyl (2S)-2-(methylamino)-3-phenylpropanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.CNC(CC1=CC=CC=C1)C(=O)OCC2=CC=CC=C2
Beilstein 6311000
Formula Weight 441.54
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Methyl-L-phenylalanine benzyl ester p-toluenesulfonate

storageNote1 : 2° to 8°C