Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-L-prolinol, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
34381-71-0
C6H13NO
115.18
VCOJPHPOVDIRJK-LURJTMIESA-N
n-methyl-l-prolinol,s---1-methyl-2-pyrrolidinemethanol,n-methylprolinol,2s-1-methylpyrrolidin-2-yl methanol,s-1-methylpyrrolidin-2-yl methanol,2-pyrrolidinemethanol, 1-methyl-, 2s,2s-1-methyl-2-pyrrolidinyl methanol,s---2-hydroxymethyl-1-methylpyrrolidine,1-methyl-2-pyrrolidinyl methanol #,n-methyl prolinol
643492
[(2S)-1-methylpyrrolidin-2-yl]methanol
CN1CCCC1CO
34381-71-0
C6H13NO
115.18
VCOJPHPOVDIRJK-LURJTMIESA-N
n-methyl-l-prolinol,s---1-methyl-2-pyrrolidinemethanol,n-methylprolinol,2s-1-methylpyrrolidin-2-yl methanol,s-1-methylpyrrolidin-2-yl methanol,2-pyrrolidinemethanol, 1-methyl-, 2s,2s-1-methyl-2-pyrrolidinyl methanol,s---2-hydroxymethyl-1-methylpyrrolidine,1-methyl-2-pyrrolidinyl methanol #,n-methyl prolinol
643492
[(2S)-1-methylpyrrolidin-2-yl]methanol
CN1CCCC1CO
Specific Gravity 0.97
Refractive Index 1.4692
Flash Point 63°C
Boiling Point 67°C to 69°C (12.0 Torr)
Density 0.9700g/mL
Color Colorless to Yellow
Quantity 5g
Specific Rotation -46° to -54° (20°C, 589nm) (c=5 in methanol)
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Solubility Information Solubility in water: in water:(20°C) insoluble.,in ethanol:(20°C) soluble
Formula Weight 115.18
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material N-Methyl-L-prolinol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 251-981-9