Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-L-valine methyl ester hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
3339-44-4
C7H16ClNO2
181.66
MFCD00238325
DLLHGHUMLSLOID-RGMNGODLSA-N
n-me-val-ome hcl,n-methyl-l-valine methyl ester hydrochloride,n-me-val-ome . hcl,h-nmeval-ome hydrochloride,methyl n-methyl-l-valinate hydrochloride,n-methyl-l-valine methyl ester hydrochlorid,l-valine, n-methyl-, methyl ester, hydrochloride,methyl 2s-3-methyl-2-methylamino butanoate hydrochloride
13819431
methyl (2S)-3-methyl-2-(methylamino)butanoate;hydrochloride
CC(C)C(C(=O)OC)NC.Cl
3339-44-4
C7H16ClNO2
181.66
MFCD00238325
DLLHGHUMLSLOID-RGMNGODLSA-N
n-me-val-ome hcl,n-methyl-l-valine methyl ester hydrochloride,n-me-val-ome . hcl,h-nmeval-ome hydrochloride,methyl n-methyl-l-valinate hydrochloride,n-methyl-l-valine methyl ester hydrochlorid,l-valine, n-methyl-, methyl ester, hydrochloride,methyl 2s-3-methyl-2-methylamino butanoate hydrochloride
13819431
methyl (2S)-3-methyl-2-(methylamino)butanoate;hydrochloride
CC(C)C(C(=O)OC)NC.Cl
Beilstein 3562296
Formula Weight 181.66
Physical Form Solid
Percent Purity 95%
Melting Point 103°C to 104°C
Quantity 5g
Chemical Name or Material N-Methyl-L-valine methyl ester hydrochloride

storageNote1 : 2° to 8°C