Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine, 97%, Alfa Aesar™

Click to view available options
Quantity:
0.25g
1g
120940-43-4
C8H8F3N3O2
235.166
MFCD00052691
GBBAZCATUSUQOJ-UHFFFAOYSA-N
1-methyl-1-2-nitro-4-trifluoromethyl phenyl hydrazine,n-methyl-n-2-nitro-4-trifluoromethyl phenyl hydrazine,hydrazine,1-methyl-1-2-nitro-4-trifluoromethyl phenyl,aminomethyl 2-nitro-4-trifluoromethyl phenyl amine,acmc-1cirx,1-methyl-1-2-nitro-4-trifluoromethyl phenyl diazane
2775458
1-methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
CN(C1=C(C=C(C=C1)C(F)(F)F)[N+](=O)[O-])N
120940-43-4
C8H8F3N3O2
235.166
MFCD00052691
GBBAZCATUSUQOJ-UHFFFAOYSA-N
1-methyl-1-2-nitro-4-trifluoromethyl phenyl hydrazine,n-methyl-n-2-nitro-4-trifluoromethyl phenyl hydrazine,hydrazine,1-methyl-1-2-nitro-4-trifluoromethyl phenyl,aminomethyl 2-nitro-4-trifluoromethyl phenyl amine,acmc-1cirx,1-methyl-1-2-nitro-4-trifluoromethyl phenyl diazane
2775458
1-methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
CN(C1=C(C=C(C=C1)C(F)(F)F)[N+](=O)[O-])N
Melting Point 64°C to 67°C
Quantity 1g
Formula Weight 235.16
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning