Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(4-pyridylmethyl)amine, 97+%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
6971-44-4
C7H10N2
122.171
DNBWGFKLIBQQSL-UHFFFAOYSA-N
n-methyl-1-pyridin-4-yl methanamine, n-methyl-n-4-pyridylmethyl amine, 4-pyridinemethanamine, n-methyl, methylpyridin-4-ylmethylamine, n-methyl-4-pyridylmethylamine, n-methyl-n-4-pyridinylmethyl amine, methyl pyridin-4-ylmethyl amine, n-methylpyridine-4-methylamine, n-methyl-n-pyridin-4-ylmethyl amine, methyl 4-pyridylmethyl amine
81436
N-methyl-1-pyridin-4-ylmethanamine
CNCC1=CC=NC=C1
6971-44-4
C7H10N2
122.171
DNBWGFKLIBQQSL-UHFFFAOYSA-N
n-methyl-1-pyridin-4-yl methanamine, n-methyl-n-4-pyridylmethyl amine, 4-pyridinemethanamine, n-methyl, methylpyridin-4-ylmethylamine, n-methyl-4-pyridylmethylamine, n-methyl-n-4-pyridinylmethyl amine, methyl pyridin-4-ylmethyl amine, n-methylpyridine-4-methylamine, n-methyl-n-pyridin-4-ylmethyl amine, methyl 4-pyridylmethyl amine
81436
N-methyl-1-pyridin-4-ylmethanamine
CNCC1=CC=NC=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(4-pyridinylmethyl)amine