Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-propargylbenzylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
555-57-7
C11H13N
159.23
MFCD00008576
DPWPWRLQFGFJFI-UHFFFAOYSA-N
pargyline,pargylamine,n-methyl-n-propargylbenzylamine,paragyline,eudatin,supirdyl,eutonyl,pargylin,n-methyl-n-2-propynylbenzylamine,benzyl-methyl-2-propinylamin
4688
CHEBI:7930
N-benzyl-N-methylprop-2-yn-1-amine
CN(CC#C)CC1=CC=CC=C1
555-57-7
C11H13N
159.23
MFCD00008576
DPWPWRLQFGFJFI-UHFFFAOYSA-N
pargyline,pargylamine,n-methyl-n-propargylbenzylamine,paragyline,eudatin,supirdyl,eutonyl,pargylin,n-methyl-n-2-propynylbenzylamine,benzyl-methyl-2-propinylamin
4688
CHEBI:7930
N-benzyl-N-methylprop-2-yn-1-amine
CN(CC#C)CC1=CC=CC=C1
Boiling Point 86°C to 88°C (4.0mmHg)
Flash Point 83°C
Density 0.9400g/mL
Specific Gravity 0.94
Refractive Index 1.5213
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH2N(CH3)CH2C∽CH
Beilstein 12, II, 548
Merck Index 15, 7142
Solubility Information Solubility in water: soluble. Other solubilities: miscibel in ethanol,acetone
Formula Weight 159.23
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material N-Methyl-N-propargylbenzylamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning