Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(trimethylsilyl)acetamide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
7449-74-3
C6H15NOSi
145.277
MFCD00040758
QHUOBLDKFGCVCG-UHFFFAOYSA-N
n-methyl-n-trimethylsilyl acetamide,acetamide, n-methyl-n-trimethylsilyl,n-trimethylsilyl-n-methylacetamide,n-methyl-n-tms-acetamide,acmc-1bei6,qhuobldkfgcvcg-uhfffaoysa,n-methylacetamide, tms derivative,n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
81953
N-methyl-N-trimethylsilylacetamide
CC(=O)N(C)[Si](C)(C)C
7449-74-3
C6H15NOSi
145.277
MFCD00040758
QHUOBLDKFGCVCG-UHFFFAOYSA-N
n-methyl-n-trimethylsilyl acetamide,acetamide, n-methyl-n-trimethylsilyl,n-trimethylsilyl-n-methylacetamide,n-methyl-n-tms-acetamide,acmc-1bei6,qhuobldkfgcvcg-uhfffaoysa,n-methylacetamide, tms derivative,n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
81953
N-methyl-N-trimethylsilylacetamide
CC(=O)N(C)[Si](C)(C)C
Silylation reagent
Color Yellow
Density 0.9g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CON(CH3)Si(CH3)3
Beilstein 04, IV, 4011
Formula Weight 145.28
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material N-Methyl-N-(trimethylsilyl)acetamide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-217-0

TSCA : TSCA