Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-p-anisidine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
5961-59-1
C8H11NO
137.18
MFCD00008399
JFXDIXYFXDOZIT-UHFFFAOYSA-N
n-methyl-p-anisidine, n-methyl-4-anisidine, n-methyl-4-methoxyaniline, benzenamine, 4-methoxy-n-methyl, n-methyl-p-ansidine, p-anisidine, n-methyl, 4-methoxy-n-methylbenzenamine, 1-methylamino-4-methoxybenzene, p-methoxy-n-methylaniline, acmc-209mes
22250
4-methoxy-N-methylaniline
CNC1=CC=C(C=C1)OC
5961-59-1
C8H11NO
137.18
MFCD00008399
JFXDIXYFXDOZIT-UHFFFAOYSA-N
n-methyl-p-anisidine, n-methyl-4-anisidine, n-methyl-4-methoxyaniline, benzenamine, 4-methoxy-n-methyl, n-methyl-p-ansidine, p-anisidine, n-methyl, 4-methoxy-n-methylbenzenamine, 1-methylamino-4-methoxybenzene, p-methoxy-n-methylaniline, acmc-209mes
22250
4-methoxy-N-methylaniline
CNC1=CC=C(C=C1)OC
Color Brown-Gray to White
Quantity 10g
Flash Point 110°C
Infrared Spectrum Authentic
Melting Point 33.0°C to 36.0°C
Packaging Glass bottle
Boiling Point 135.0°C to 136.0°C (19.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4NHCH3
Beilstein 13, 442
Formula Weight 137.18
Physical Form Crystalline Low Melting Mass
Percent Purity 98%
Chemical Name or Material N-Methyl-p-anisidine, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/va

GHS Signal Word: Warning

EINECSNumber : 227-735-1