Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methylacetamide, 99%

Click to view available options
Quantity:
250g
2500g
10000g
79-16-3
C3H7NO
73.095
MFCD00008683
OHLUUHNLEMFGTQ-UHFFFAOYSA-N
acetamide, n-methyl,methylacetamide,monomethylacetamide,n-acetyl-n-methylamine,acetylmethylamine,methyl acetamide,n-methyl acetamide,n-methyl-acetamide,acetamide, methyl,n-monomethylacetamide
6582
CHEBI:87321
N-methylacetamide
CC(=O)NC
79-16-3
C3H7NO
73.095
MFCD00008683
OHLUUHNLEMFGTQ-UHFFFAOYSA-N
acetamide, n-methyl,methylacetamide,monomethylacetamide,n-acetyl-n-methylamine,acetylmethylamine,methyl acetamide,n-methyl acetamide,n-methyl-acetamide,acetamide, methyl,n-monomethylacetamide
6582
CHEBI:87321
N-methylacetamide
CC(=O)NC
Applications
N-Methylacetamide is used as a chemical intermediate in the production of life science, agrochemicals, electronic materials and construction materials. It is also used as a solvent, in electrochemistry.

Solubility
Soluble in water (1X10+6 mg/L @ 20 deg C), chloroform. Soluble in alcohol, ether, acetone, benzene.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Odor Odorless
Density 0.957
Refractive Index 1.433
Melting Point 26°C to 28°C
Boiling Point 204°C to 206°C
Quantity 2500g
Flash Point 116°C (240°F)
Beilstein 1071255
Solubility Information Soluble in water (1X10+6mg/L @ 20 deg C),chloroform. Soluble in alcohol,ether,acetone,benzene.
Formula Weight 73.1
Percent Purity 99%
Chemical Name or Material N-Methylacetamide
Show More Show Less

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360D

EINECSNumber : 201-182-6

RTECSNumber : AC5960000

TSCA : Yes

Recommended Storage : Ambient temperatures