Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylacetamide, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
79-16-3
C3H7NO
73.095
MFCD00008683
OHLUUHNLEMFGTQ-UHFFFAOYSA-N
acetamide, n-methyl,methylacetamide,monomethylacetamide,n-acetyl-n-methylamine,acetylmethylamine,methyl acetamide,n-methyl acetamide,n-methyl-acetamide,acetamide, methyl,n-monomethylacetamide
6582
CHEBI:87321
N-methylacetamide
CC(=O)NC
79-16-3
C3H7NO
73.095
MFCD00008683
OHLUUHNLEMFGTQ-UHFFFAOYSA-N
acetamide, n-methyl,methylacetamide,monomethylacetamide,n-acetyl-n-methylamine,acetylmethylamine,methyl acetamide,n-methyl acetamide,n-methyl-acetamide,acetamide, methyl,n-monomethylacetamide
6582
CHEBI:87321
N-methylacetamide
CC(=O)NC
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Density 0.946g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3CONHCH3
Beilstein 04, 58
Formula Weight 73.1
Physical Form Liquid After Melting
Percent Purity ≥99.0%
Chemical Name or Material N-Methylacetamide, 99+%
Show More Show Less

GHS H Statement:
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-182-6

RTECSNumber : AC5960000

TSCA : TSCA