Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
Packaging:
Glass bottle
100-61-8
C7H9N
107.15
MFCD00008283
AFBPFSWMIHJQDM-UHFFFAOYSA-N
methylaniline,monomethylaniline,n-methyl aniline,benzenamine, n-methyl,methylphenylamine,n-methylbenzenamine,n-monomethylaniline,n-methylaminobenzene,methylamino benzene,anilinomethane
7515
CHEBI:15733
N-methylaniline
CNC1=CC=CC=C1
100-61-8
C7H9N
107.15
MFCD00008283
AFBPFSWMIHJQDM-UHFFFAOYSA-N
methylaniline,monomethylaniline,n-methyl aniline,benzenamine, n-methyl,methylphenylamine,n-methylbenzenamine,n-monomethylaniline,n-methylaminobenzene,methylamino benzene,anilinomethane
7515
CHEBI:15733
N-methylaniline
CNC1=CC=CC=C1
Packaging Glass bottle
Melting Point -57.0°C
Refractive Index 1.5694 to 1.5714
Specific Gravity 0.987
Boiling Point 195.0°C
Infrared Spectrum Authentic
Quantity 500g
Density 0.9870g/mL
Color Brown to Yellow
Flash Point 86°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5NHCH3
Beilstein 12, 135
Fieser 14,207
Merck Index 15, 6093
Solubility Information Solubility in water: 30g/l. Other solubilities: soluble in alcohol and ether,soluble in acetone,diethyl ether,ethanol and,chloroform
Formula Weight 107.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Methylaniline
Show More Show Less

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger