Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Methyldi-n-propylamine, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N-Methyldi-n-propylamine can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in hot water.
Notes
Do not store together with acids. Incompatible with acids and oxidizing agents.
N-Methyldi-n-propylamine can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in hot water.
Notes
Do not store together with acids. Incompatible with acids and oxidizing agents.
Specifications
Color | Colorless |
Density | 0.743g/mL |
Boiling Point | 113°C to 117°C |
Refractive Index | 1.4060 |
Linear Formula | (CH3CH2CH2)2NCH3 |
Quantity | 25 g |
UN Number | UN2924 |
Beilstein | 1731704 |
Formula Weight | 115.22 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | GHS P Statement P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction