Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyldi-n-propylamine, 98%

Click to view available options
Quantity:
5g
25g
3405-42-3
C7H17N
115.22
MFCD00048683
UVBMZKBIZUWTLV-UHFFFAOYSA-N
n-methyldipropylamine,n-methyl-n-propyl-propylamine,methyldipropylamine,1-propanamine, n-methyl-n-propyl,methyl di-n-propylamine,methyl dipropyl amine,n-methyl dipropylamine,n-methyldipropyl amine,n,n-dipropylmethylamine,methyl di-n-propyl amine
559387
N-methyl-N-propylpropan-1-amine
CCCN(C)CCC
3405-42-3
C7H17N
115.22
MFCD00048683
UVBMZKBIZUWTLV-UHFFFAOYSA-N
n-methyldipropylamine,n-methyl-n-propyl-propylamine,methyldipropylamine,1-propanamine, n-methyl-n-propyl,methyl di-n-propylamine,methyl dipropyl amine,n-methyl dipropylamine,n-methyldipropyl amine,n,n-dipropylmethylamine,methyl di-n-propyl amine
559387
N-methyl-N-propylpropan-1-amine
CCCN(C)CCC
Applications
N-Methyldi-n-propylamine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot water.

Notes
Do not store together with acids. Incompatible with acids and oxidizing agents.
Color Colorless
Density 0.743g/mL
Boiling Point 113°C to 117°C
Quantity 5g
Refractive Index 1.4060
Linear Formula (CH3CH2CH2)2NCH3
UN Number UN2924
Beilstein 1731704
Formula Weight 115.22
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material N-Methyldi-n-propylamine
Show More Show Less

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.