Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyldioctylamine, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
4455-26-9
C17H37N
255.49
MFCD00009559
YJLYANLCNIKXMG-UHFFFAOYSA-N
methyldioctylamine,n-methyldioctylamine,n-methyldi-n-octylamine,di octyl methylamine,n,n-dioctylmethylamine,1-octanamine, n-methyl-n-octyl,di-n-octylmethylamine,n,n-dioctyl-n-methylamine,n-methyl-n-octyl-1-octanamine,dioctyl methyl amine
78202
N-methyl-N-octyloctan-1-amine
CCCCCCCCN(C)CCCCCCCC
4455-26-9
C17H37N
255.49
MFCD00009559
YJLYANLCNIKXMG-UHFFFAOYSA-N
methyldioctylamine,n-methyldioctylamine,n-methyldi-n-octylamine,di octyl methylamine,n,n-dioctylmethylamine,1-octanamine, n-methyl-n-octyl,di-n-octylmethylamine,n,n-dioctyl-n-methylamine,n-methyl-n-octyl-1-octanamine,dioctyl methyl amine
78202
N-methyl-N-octyloctan-1-amine
CCCCCCCCN(C)CCCCCCCC
Infrared Spectrum Authentic
Quantity 5g
Color Colorless to Yellow
Packaging Glass bottle
Refractive Index 1.4414 to 1.4434 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula {CH3(CH2)7}2NCH3
Beilstein 04, III, 381
Formula Weight 255.49
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material N-Methyldioctylamine, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.