Alfa Aesar™ N-Methyldiphenylamine, 98%

Manufacturer: Alfa Aesar™ B2473906

Quantity 5g

 View more versions of this product

Catalog No. AAB2473906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 552-82-9
    Molecular Formula (C6H5)2NCH3
    Formula Weight 183.25
    MDL Number MFCD00041900
    Merck Index 14,6054
    Synonym N,N-Diphenylmethylamine
    Chemical Name or Material N-Methyldiphenylamine

    Specifications

    Formula Weight 183.25
    TSCA TSCA
    Quantity 5g
    Flash Point >110°C (230°F)
    Molecular Formula (C6H5)2NCH3
    CAS 552-82-9
    Merck Index 14,6054
    Health Hazard 1 Warning
    Boiling Point 296° to 297°C
    MDL Number MFCD00041900
    Synonym N,N-Diphenylmethylamine
    Solubility Information Insoluble in water; Soluble in alcohol, ether
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material N-Methyldiphenylamine
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit