Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylethylenediamine, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
109-81-9
C3H10N2
74.13
MFCD00008165
KFIGICHILYTCJF-UHFFFAOYSA-N
n-methylethylenediamine,1,2-ethanediamine, n-methyl,2-aminoethylmethylamine,n-methyldiaminoethane,n-methylethanediamine,2-methylamino ethylamine,n-methyl-1,2-ethanediamine,n-methylethylidenediamine,ethylenediamine, n-methyl,n-methylethane-1,2-diamine
8014
N'-methylethane-1,2-diamine
CNCCN
109-81-9
C3H10N2
74.13
MFCD00008165
KFIGICHILYTCJF-UHFFFAOYSA-N
n-methylethylenediamine,1,2-ethanediamine, n-methyl,2-aminoethylmethylamine,n-methyldiaminoethane,n-methylethanediamine,2-methylamino ethylamine,n-methyl-1,2-ethanediamine,n-methylethylidenediamine,ethylenediamine, n-methyl,n-methylethane-1,2-diamine
8014
N'-methylethane-1,2-diamine
CNCCN
Refractive Index 1.4390 to 1.4420
Specific Gravity 0.84
Flash Point 41°C
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Boiling Point 114.0°C to 116.0°C
Density 0.8400g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3NHCH2CH2NH2
Beilstein 04, I, 415
Formula Weight 74.13
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material N-Methylethylenediamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-707-4