Alfa Aesar™ N-Methylethylenediamine, 98%

Manufacturer: Alfa Aesar™ 3186604

Quantity 2g

 View more versions of this product

Catalog No. AA3186604

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 109-81-9
    Formula Weight 74.13
    MDL Number MFCD00008165
    UN Number UN2733
    Synonym 2-Methylaminoethylamine
    Chemical Name or Material N-Methylethylenediamine

    Specifications

    Formula Weight 74.13
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Refractive Index 1.4395
    Density 0.841g/mL
    Odor Ammonia-like
    UN Number UN2733
    Synonym 2-Methylaminoethylamine
    Boiling Point 114° to 116°C
    Physical Form Liquid
    Health Hazard 1 Danger
    TSCA TSCA
    Quantity 2g
    Flash Point 41° (106°F)
    MDL Number MFCD00008165
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    CAS 109-81-9
    Chemical Name or Material N-Methylethylenediamine
    Assay 98%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit