Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylethylenediamine 97.0+%, TCI America™
SDP

Catalog No. p-7132213
Click to view available options
Quantity:
5 mL
25 mL
100 mL

Chemical Identifiers

CAS 109-81-9
Molecular Formula C3H12N2
Molecular Weight (g/mol) 76.14
MDL Number MFCD00008165
InChI Key KFIGICHILYTCJF-UHFFFAOYSA-P
Synonym n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
PubChem CID 8014
IUPAC Name (2-azaniumylethyl)(methyl)azanium
SMILES C[NH2+]CC[NH3+]

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 mL
UN Number 2734
Formula Weight 74.13
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material N-Methylethylenediamine
RTECSNumber KV5250500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.