Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylformanilide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
1kg
10kg
Packaging:
Glass bottle
Plastic drum
93-61-8
C8H9NO
135.17
MFCD00003283
JIKUXBYRTXDNIY-UHFFFAOYSA-N
n-methylformanilide,formamide, n-methyl-n-phenyl,n-formyl-n-methylaniline,formanilide, n-methyl,methylphenylformamide,n-phenyl-n-methylformamide,n-methyl-n-formylaniline,n-methylformanilid,methyl phenyl formamide,n-methyl-n-phenyl-formamide
66737
N-methyl-N-phenylformamide
CN(C=O)C1=CC=CC=C1
93-61-8
C8H9NO
135.17
MFCD00003283
JIKUXBYRTXDNIY-UHFFFAOYSA-N
n-methylformanilide,formamide, n-methyl-n-phenyl,n-formyl-n-methylaniline,formanilide, n-methyl,methylphenylformamide,n-phenyl-n-methylformamide,n-methyl-n-formylaniline,n-methylformanilid,methyl phenyl formamide,n-methyl-n-phenyl-formamide
66737
N-methyl-N-phenylformamide
CN(C=O)C1=CC=CC=C1
Melting Point 8.0°C to 13.0°C
Boiling Point 243.0°C to 244.0°C
Specific Gravity 1.09
Refractive Index 1.5600 to 1.5620
Infrared Spectrum Authentic
Flash Point 126°C
Packaging Glass bottle
Quantity 500g
Color Colorless to Yellow
Density 1.0900g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HCON(CH3)C6H5
Beilstein 12,234
Fieser 01,68
Solubility Information Solubility in water: immiscible.
Formula Weight 135.17
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Methylformanilide, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

EINECSNumber : 202-262-3

TSCA : TSCA