Nα-Methylhistamine dihydrochloride, Tocris

Non-selective H3 agonist

Manufacturer: R&D Systems 0573/50

Quantity 50mg

 View more versions of this product

Catalog No. 05-735-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C6H11N3·2HCl
    Formula Weight 198.09
    Synonym N-Methyl-1H-imidazole-4-ethanamine dihydrochloride
    Chemical Name or Material Nα-Methylhistamine dihydrochloride

    Specifications

    Quantity 50mg
    Formula Weight 198.09
    Assay Percent Range >98%
    Molecular Formula C6H11N3·2HCl
    CAS 16503-22-3
    Synonym N-Methyl-1H-imidazole-4-ethanamine dihydrochloride
    Recommended Storage Store at Room Temperature
    Chemical Name or Material Nα-Methylhistamine dihydrochloride

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit