Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyliminodiacetic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4408-64-4
C5H9NO4
147.13
MFCD00004284
XWSGEVNYFYKXCP-UHFFFAOYSA-N
n-methyliminodiacetic acid,2,2'-methylazanediyl diacetic acid,methyliminodiacetic acid,mida,methylimidodiacetic acid,n-methyliminodiaceticacid,glycine, n-carboxymethyl-n-methyl,methylimino diacetic acid,acetic acid, methylimino di,carboxymethyl methyl amino acetic acid
20441
2-[carboxymethyl(methyl)amino]acetic acid
CN(CC(=O)O)CC(=O)O
4408-64-4
C5H9NO4
147.13
MFCD00004284
XWSGEVNYFYKXCP-UHFFFAOYSA-N
n-methyliminodiacetic acid,2,2'-methylazanediyl diacetic acid,methyliminodiacetic acid,mida,methylimidodiacetic acid,n-methyliminodiaceticacid,glycine, n-carboxymethyl-n-methyl,methylimino diacetic acid,acetic acid, methylimino di,carboxymethyl methyl amino acetic acid
20441
2-[carboxymethyl(methyl)amino]acetic acid
CN(CC(=O)O)CC(=O)O
Melting Point 218°C to 221°C
Infrared Spectrum Authentic
Color Cream to White
Quantity 25g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3N(CH2CO2H)2
Beilstein 04, 367
Formula Weight 147.13
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material N-Methyliminodiacetic acid, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 224-557-6