Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylindole-3-boronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
683229-61-0
C15H20BNO2
257.14
MCCCOWZSGBKRCG-UHFFFAOYSA-N
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole,n-methylindole-3-boronic acid pinacol ester,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole,n-methylindole-3-boronic acid, pinacol ester,1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole,1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl indole,1-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-1h-indole,1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
23058003
1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
B1(OC(C(O1)(C)C)(C)C)C2=CN(C3=CC=CC=C23)C
683229-61-0
C15H20BNO2
257.14
MCCCOWZSGBKRCG-UHFFFAOYSA-N
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole,n-methylindole-3-boronic acid pinacol ester,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole,n-methylindole-3-boronic acid, pinacol ester,1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole,1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl indole,1-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole,1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-1h-indole,1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
23058003
1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
B1(OC(C(O1)(C)C)(C)C)C2=CN(C3=CC=CC=C23)C
Melting Point 106°C to 112°C
Quantity 5g
Infrared Spectrum Authentic
Color Brown to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 257.14
Physical Form Low Melting Solid
Percent Purity 97%
Chemical Name or Material N-Methylindole-3-boronic Acid Pinacol Ester
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning