Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmaleimide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
930-88-1
C5H5NO2
111.1
MFCD00005508
SEEYREPSKCQBBF-UHFFFAOYSA-N
n-methylmaleimide,1-methyl-1h-pyrrole-2,5-dione,maleimide, n-methyl,n-methylmaleinimide,1h-pyrrole-2,5-dione, 1-methyl,n-methyl maleimide,unii-p0tfz8r21y,p0tfz8r21y,1-methyl-pyrrole-2,5-dione,1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione
70261
1-methylpyrrole-2,5-dione
CN1C(=O)C=CC1=O
930-88-1
C5H5NO2
111.1
MFCD00005508
SEEYREPSKCQBBF-UHFFFAOYSA-N
n-methylmaleimide,1-methyl-1h-pyrrole-2,5-dione,maleimide, n-methyl,n-methylmaleinimide,1h-pyrrole-2,5-dione, 1-methyl,n-methyl maleimide,unii-p0tfz8r21y,p0tfz8r21y,1-methyl-pyrrole-2,5-dione,1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione
70261
1-methylpyrrole-2,5-dione
CN1C(=O)C=CC1=O
Quantity 5g
Color Yellow
Melting Point 93.0°C to 98.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 21, 399
Formula Weight 111.1
Physical Form Crystals
Percent Purity 97%
Chemical Name or Material N-Methylmaleimide
Show More Show Less

GHS H Statement:
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger