Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylphenethylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
589-08-2
C9H13N
135.21
MFCD00008291
SASNBVQSOZSTPD-UHFFFAOYSA-N
n-methylphenethylamine,benzeneethanamine, n-methyl,methyl-phenethyl-amine,n-methyl-2-phenylethylamine,n-methyl-n-2-phenylethyl amine,1-phenyl-2-methylaminoethane,phenethylamine, n-methyl,methyl 2-phenylethyl amine,1-phenyl-2-methylamino-aethan,n-methyl-phenethylamine
11503
N-methyl-2-phenylethanamine
CNCCC1=CC=CC=C1
589-08-2
C9H13N
135.21
MFCD00008291
SASNBVQSOZSTPD-UHFFFAOYSA-N
n-methylphenethylamine,benzeneethanamine, n-methyl,methyl-phenethyl-amine,n-methyl-2-phenylethylamine,n-methyl-n-2-phenylethyl amine,1-phenyl-2-methylaminoethane,phenethylamine, n-methyl,methyl 2-phenylethyl amine,1-phenyl-2-methylamino-aethan,n-methyl-phenethylamine
11503
N-methyl-2-phenylethanamine
CNCCC1=CC=CC=C1
Specific Gravity 0.93
Quantity 5g
Flash Point 73°C
Boiling Point 203.0°C
Color Colorless to Yellow
Density 0.9300g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5150 to 1.5170
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH2CH2NHCH3
Beilstein 12, 1097
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in alcohol and ether
Formula Weight 135.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Methylphenethylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger